accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHS

CAS RN 6165-68-0

IUPAC Name: (thiophen-2-yl)boronic acid
Synonyms: thiophen-2-ylboronic acid
Molecular Weight (g/mol): 127.95
Molecular Formula: C4H5BO2S
InChi Key: ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CS1
Molecular Weight (g/mol) 
  • (7)
  • (2)
  • (2)
Quantity 
  • (4)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (7)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (3)
  • (6)
  • (3)
  • (3)