Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T17725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1772-5G 5 g
T1772-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1772-5G Supplier TCI America Supplier No. T17725G
Add to Cart
Add to Cart

Specifications

CAS 6165-68-0
Color White-Yellow
Molecular Formula C4H5BO2S
MDL Number MFCD00151850
Quantity 5 g
Synonym thiophene-2-boronic acid, 2-thiopheneboronic acid, 2-thienylboronic acid, thienylboronic acid, 2-thiopheneboronicacid
InChI Key ARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILES B(C1=CC=CS1)(O)O
IUPAC Name thiophen-2-ylboronic acid
Molecular Weight (g/mol) 127.952
PubChem CID 2733960
Formula Weight 127.95
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.