accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOHHOHO

CAS RN 617-04-9

IUPAC Name: 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms: methyl-α-D-mannoside methyl mannoside mannoside, methyl
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H14O6
InChi Key: HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES: COC1OC(CO)C(O)C(O)C1O
Quantity 
  • (2)
  • (4)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (9)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (4)
  • (4)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (9)
  • (3)
  • (4)