Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl α-D-Mannopyranoside, MP Biomedicals™

Catalog No. MP215393790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215393790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215393790 Supplier MP Biomedicals, Inc Supplier No. 0215393790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reduces Con A binding to the lymphocyte surface in culture. Methyl α-D-Mannopyranoside, MP Biomedicals a competitor inhibitor of the binding of mannose by Escherichia coli; it was used in a study to investigate the primary mannose binding site of pr

Methyl α-D-mannopyranoside has been used to synthesize a series of tri- and tetrahydroxylated seven-membered iminosugars in a study that worked towards a stable noeuromycin analog with a D-manno configuration.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 187°C to 197°C
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 194.20
Percent Purity ∼99%
Grade Not Available
Physical Form Crystalline Powder
Recommended Storage Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.