accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONH2H3CHCl

CAS RN 617-27-6

IUPAC Name: ethyl 2-aminopropanoate hydrochloride
Synonyms: ethyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: JCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C(C)N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results