Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AC400430250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC400430250 Supplier Thermo Scientific Chemicals Supplier No. 400430250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 617-27-6 | Molecular Formula: C5H12ClNO2 | Molecular Weight: 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-27-6
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00013018
InChI Key JCXLZWMDXJFOOI-UHFFFAOYNA-N
Synonym ethyl 2-aminopropanoate hydrochloride, dl-alanine ethyl ester hydrochloride, h-dl-ala-oet.hcl, h-dl-ala-oet hydrochloride, d,l-alanine ethyl ester hydrochloride, h-dl-ala-oet hcl, l-alanine ethyl ester hcl, alanine, ethyl ester, hydrochloride 1:1, alanine, ethyl ester, hydrochloride, ethyl alaninate hydrochloride
PubChem CID 102560
IUPAC Name ethyl 2-aminopropanoate;hydrochloride
SMILES Cl.CCOC(=O)C(C)N

Specifications

Melting Point 87.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH(NH)2COOC2H5·HCl
Quantity 25 g
Formula Weight 153.61
Percent Purity 99%
Physical Form Granular Powder and Granules
Chemical Name or Material DL-Alanine ethyl ester hydrochloride, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-507-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.