accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 617-52-7

IUPAC Name: 1,4-dimethyl 2-methylidenebutanedioate
Synonyms: 1,4-dimethyl 2-methylidenebutanedioate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: ZWWQRMFIZFPUAA-UHFFFAOYSA-N
SMILES: COC(=O)CC(=C)C(=O)OC
Boiling Point 
  • (2)
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
  • (2)
Quantity 
  • (4)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)