Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl itaconate

Catalog No. 109533500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-953-3500G 500 g
10-953-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-953-3500G Supplier Sigma Aldrich Supplier No. 109533500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl itaconate

Specifications

CAS 617-52-7
Melting Point 37°C to 41°C (lit.)
Density 1.124 g/mL (at 25°C (literature))
Boiling Point 208°C (lit.)
Molecular Formula C7H10O4
Linear Formula CH3O2CCH2C(=CH2)CO2CH3
MDL Number MFCD00008446
Quantity 500 g
Molecular Weight (g/mol) 158.15
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.