accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 617-78-7

IUPAC Name: 3-ethylpentane
Synonyms: 3-ethylpentane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: AORMDLNPRGXHHL-UHFFFAOYSA-N
SMILES: CCC(CC)CC
Percent Purity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results