Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylpentane 93.0+%, TCI America™
SDP

Catalog No. E00465ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00465ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E00465ML Supplier TCI America Supplier No. E00465ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 617-78-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00027083
InChI Key AORMDLNPRGXHHL-UHFFFAOYSA-N
Synonym Triethylmethane
PubChem CID 12048
IUPAC Name 3-ethylpentane
SMILES CCC(CC)CC

Specifications

Physical Form Liquid
UN Number 3295
Color Colorless
Boiling Point 94°C
Quantity 5 mL
Formula Weight 100.21
Percent Purity ≥93.0% (GC)
Chemical Name or Material 3-Ethylpentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.