accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CO

CAS RN 617-84-5

IUPAC Name: N,N-diethylformamide
Synonyms: N,N-diethylformamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: SUAKHGWARZSWIH-UHFFFAOYSA-N
SMILES: CCN(CC)C=O
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (7)
  • (9)