Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Diethylformamide

Catalog No. 18631725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-631-725G 25 g
18-631-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-631-725G Supplier Sigma Aldrich Supplier No. 18631725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-diethylformamide undergoes condensation with aromatic o-aminonitriles in the presence of zncl2 in sealed reactor to yield quinazoline-2,4(1h,3h)-dione.n,n-diethylformamide was used in the synthesis of metal-organic frameworks
  • It was also used as solvent in the synthesis of porous cubic-shaped zno particles

Specifications

CAS 617-84-5
Density 0.908 g/mL (at 25°C (literature))
Boiling Point 176°C to 177°C (lit.)
Molecular Formula C5H11NO
Linear Formula HCON(C2H5)2
Refractive Index n20/D 1.434 (literature)
MDL Number MFCD00003287
Quantity 25 g
Molecular Weight (g/mol) 101.15
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.