accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOOOH

CAS RN 6174-86-3

IUPAC Name: 3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one
Synonyms: chlorferone chlorferron C10H7ClO3
Molecular Weight (g/mol): 210.61
Molecular Formula: C10H7ClO3
InChi Key: ODZHLDRQCZXQFQ-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)