Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-7-hydroxy-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. C30985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30985G 5 g
C30981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30985G Supplier TCI America Supplier No. C30985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6174-86-3
Molecular Formula C10H7ClO3
Molecular Weight (g/mol) 210.61
MDL Number MFCD00006851
InChI Key ODZHLDRQCZXQFQ-UHFFFAOYSA-N
Synonym chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl
PubChem CID 5355079
ChEBI CHEBI:38620
IUPAC Name 3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one
SMILES CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12

Specifications

Melting Point 243°C
Quantity 5 g
Formula Weight 210.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-7-hydroxy-4-methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.