accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61748-86-5

IUPAC Name: 3,3,4-trimethylpyrrolidine-2,5-dione
Synonyms: 3,3,4-trimethylpyrrolidine-2,5-dione
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: SUPYQTKKYLUOBW-UHFFFAOYNA-N
SMILES: CC1C(=O)NC(=O)C1(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results