Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Dimethyl-β-methylsuccinimide, 99+%, Thermo Scientific™

Catalog No. AC172510010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172510010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC172510010 Supplier Thermo Scientific Chemicals Supplier No. 172510010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61748-86-5
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00005497
InChI Key SUPYQTKKYLUOBW-UHFFFAOYSA-N
Synonym 2,2,3-trimethylsuccinimide, alpha,alpha-dimethyl-beta-methylsuccinimide, a,a-dimethyl-b-methylsuccinimide, supyqtkkyluobw-uhfffaoysa, 3,3,4-trimethylazolidine-2,5-dione, 2,5-pyrrolidinedione, 3,3,4-trimethyl, 3,3,4-trimethyl-2,5-pyrrolidinedione #, .alpha.,.alpha.-dimethyl-.beta.-methylsuccinimide
PubChem CID 550809
IUPAC Name 3,3,4-trimethylpyrrolidine-2,5-dione
SMILES CC1C(=O)NC(=O)C1(C)C

Specifications

Melting Point 117°C to 122°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 1 g
Beilstein 21, 394
Formula Weight 141.17
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material α, α-Dimethyl-β-methylsuccinimide
EINECSNumber 262-956-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.