accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6178-44-5

IUPAC Name: ethyl 3,4,5-trimethoxybenzoate
Synonyms: ethyl 3,4,5-trimethoxybenzoate
Molecular Weight (g/mol): 240.26
Molecular Formula: C12H16O5
InChi Key: UEOFNBCUGJADBM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(OC)=C(OC)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results