Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3,4,5-trimethoxybenzoate, 98%

Catalog No. AAA1319922
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319922 100 g
AAA1319906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1319922 Supplier Thermo Scientific Chemicals Supplier No. A1319922
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6178-44-5 | C12H16O5 | 240.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6178-44-5
Molecular Formula C12H16O5
Molecular Weight (g/mol) 240.255
MDL Number MFCD00017525
InChI Key UEOFNBCUGJADBM-UHFFFAOYSA-N
PubChem CID 231162
IUPAC Name ethyl 3,4,5-trimethoxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)OC)OC)OC

Specifications

Quantity 100 g
Synonym 3,4,5-trimethoxybenzoic acid ethyl ester, ethyl 3,4,5-trimethoxy benzoate, ethyltrimethoxy benzoate, acmc-1b7mp, ethyl 3,4,5-trimethoxybenzoate;, benzoic acid,3,4,5-trimethoxy-, ethyl ester, benzoic acid, 3,4,5-trimethoxy-, ethyl ester
Formula Weight 240.26
Percent Purity 0.98
Chemical Name or Material Ethyl 3,4,5-trimethoxybenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.