accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClOCl

CAS RN 61785-35-1

IUPAC Name: 4-chloro-2-(2-chlorobenzoyl)phenol
Synonyms: 4-chloro-2-(2-chlorobenzoyl)phenol
Molecular Weight (g/mol): 267.11
Molecular Formula: C13H8Cl2O2
InChi Key: OSXVZDOVYCTYCW-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results