Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5-Dichloro-2-hydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. D47035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47035G 5 g
D47031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47035G Supplier TCI America Supplier No. D47035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61785-35-1
Molecular Formula C13H8Cl2O2
Molecular Weight (g/mol) 267.105
MDL Number MFCD02682994
InChI Key OSXVZDOVYCTYCW-UHFFFAOYSA-N
PubChem CID 2735995
IUPAC Name (5-chloro-2-hydroxyphenyl)-(2-chlorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)O)Cl

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 5 g
Formula Weight 267.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,5-Dichloro-2-hydroxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.