accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61788-71-4

IUPAC Name: nickel(2+) bis(naphthalene-2-carboxylate)
Synonyms: nickel(2+) bis(2-naphthoate)
Molecular Weight (g/mol): 401.04
Molecular Formula: C22H14NiO4
InChi Key: UIEKYBOPAVTZKW-UHFFFAOYSA-L
SMILES: [Ni++].[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results