Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel naphthenate, Ni 5-12%

Catalog No. AA39455A1
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39455A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39455A1 Supplier Thermo Scientific Chemicals Supplier No. 039455A1
  • $779.00
    $623.65 / Each of 1

    Save $155.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 61788-71-4 | C22H14NiO4 | 401.043 g/mol

Applied as a rubber catalyst. The product is a aid siccative and also acts as a stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61788-71-4
Molecular Formula C22H14NiO4
Molecular Weight (g/mol) 401.043
MDL Number MFCD00147723
InChI Key UIEKYBOPAVTZKW-UHFFFAOYSA-L
Synonym nickel naphthenate, naphthenic acids, nickel salts, nickel 2+ bis 2-naphthoate, nickel ii naphthenate, approx in toluene ni
PubChem CID 14647680
IUPAC Name naphthalene-2-carboxylate;nickel(2+)
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].[Ni+2]

Specifications

Color Green
Physical Form Viscous Liquid
Quantity 1 kg
UN Number UN3082
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Ni 5 to 12%
Chemical Name or Material Nickel naphthenate
Hazard Category H317-H350
Hazard Statement GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 263-000-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.