accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COO

CAS RN 61790-13-4

IUPAC Name: sodium 3-(3-ethylcyclopentyl)propanoate
Synonyms: sodium 3-(3-ethylcyclopentyl)propanoate
Molecular Weight (g/mol): 192.23
Molecular Formula: C10H17NaO2
InChi Key: KQSJSRIUULBTSE-UHFFFAOYNA-M
SMILES: [Na+].CCC1CCC(CCC([O-])=O)C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results