Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Naphthenate, TCI America™
SDP

Catalog No. N039725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. N039725G Supplier TCI America Supplier No. N039725G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 61790-13-4
Molecular Formula C10H17NaO2
Molecular Weight (g/mol) 192.234
MDL Number MFCD00147698
InChI Key KQSJSRIUULBTSE-UHFFFAOYSA-M
Synonym Naphthenic Acid Sodium Salt
PubChem CID 23687326
IUPAC Name sodium;3-(3-ethylcyclopentyl)propanoate
SMILES CCC1CCC(C1)CCC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3077
Physical Form Crystalline Powder
Chemical Name or Material Sodium Naphthenate
EINECSNumber (9)-1758
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.