CAS RN 61790-53-2
Celite™, for analysis, high purity analytical grade
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00011232 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Infusorial earth,Diatomite,Diatomaceous earth PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Diatomaceous Earth Acid-Washed, MP Biomedicals
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: silica,silicon dioxide,quartz,cristobalite,diatomaceous earth,tridymite,silicic anhydride,aerosil,infusorial earth,silica, amorphous PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Diatomaceous Earth, acid-washed, ∽95% SiO2, MP Biomedicals™
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Diatomaceous silica PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Filter aid, Celite Standard Super-cel
Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00132803 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Kieselguhr
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: silica,silicon dioxide,quartz,cristobalite,diatomaceous earth,tridymite,silicic anhydride,aerosil,infusorial earth,silica, amorphous PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Filter Aid, Celite Standard Supercel∣r
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00132803 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Celite 521AW, filter aid PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Diatomaceous Earth, Non-washed, ∽90% SiO2, MP Biomedicals™
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Diatomaceous silica PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Infusorial Earth (Kieselguhr) (Diatomaceous Silica), Reagents
CAS: 61790-53-2 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Kieselguhr, Amorphous silicon dioxide IUPAC Name: silanedione SMILES: O=[Si]=O
Volatile Residue - WP, Proficiency Testing Sample, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.