Learn More
Filter Aid, Celite Standard Supercel∣r
Description
Filter Aid, Celite AW Standard Supercel™ is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter Aid, Celite AW Standard Supercel« is used for general filtrations. It is also used as a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals,
Specifications
Specifications
| Quantity | 500 g |
| Odor | Odorless |
| Merck Index | 14,4973 |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Filter Aid, Celite AW Standard Supercel™ |
Safety and Handling
| Hazard Category | H319-H335-H373 |
| Hazard Statement | GHS H Statement H350-H373-H319-H335 May cause cancer. May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c |
| EINECSNumber | 319-127-6 |
| RTECSNumber | VV7311000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.