accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)NH(S)OHO

CAS RN 618-27-9

IUPAC Name: (2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: L-hydroxyproline 4-cis-L-hydroxyproline L-allo-hydroxyproline allo-4-hydroxyproline allo-hydroxy-L-proline cis-hydroxyproline L-allohydroxyproline L-proline, allo-hydroxy- allo-4-hydroxy-L-proline hydroxyproline cis-form
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: PMMYEEVYMWASQN-IMJSIDKUSA-N
SMILES: O[C@@H]1CN[C@@H](C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)