accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 618-31-5

IUPAC Name: (dibromomethyl)benzene
Synonyms: (dibromomethyl)benzene
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: VCJZTATVUDMNLU-UHFFFAOYSA-N
SMILES: BrC(Br)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results