Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Dibromotoluene 97%, Thermo Scientific™

Catalog No. AC154041000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC154041000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC154041000 Supplier Thermo Scientific Chemicals Supplier No. 154041000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 618-31-5
CAS Min % 96.0
CAS Max % 100.0
Density 1.51g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H6Br2
Refractive Index 1.6140 to 1.6160 (20°C, 589nm)
Linear Formula C6H5CHBr2
Quantity 100g
Beilstein 05, 308
Synonym dibromomethyl benzene, benzal bromide, alpha,alpha-dibromotoluene, benzene, dibromomethyl, benzalbromid, benzal dibromide, a,a-dibromotoluene, benzylidene bromide, benzylidene dibromide
InChI Key VCJZTATVUDMNLU-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(Br)Br
IUPAC Name dibromomethylbenzene
Molecular Weight (g/mol) 249.933
PubChem CID 69249
Formula Weight 249.93
Percent Purity ≥96.0%
Chemical Name or Material α, α-Dibromotoluene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It
EINECSNumber 210-543-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.