accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 618-42-8

IUPAC Name: 1-(piperidin-1-yl)ethan-1-one
Synonyms: 1-acetylpiperidine piperidide acetic acid acetic acid, piperidide
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: KDISMIMTGUMORD-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results