Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylpiperidine 98.0+%, TCI America™
SDP

Catalog No. A010925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A010925ML Supplier TCI America Supplier No. A010925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 618-42-8
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00006486
InChI Key KDISMIMTGUMORD-UHFFFAOYSA-N
PubChem CID 12058
ChEBI CHEBI:36591
IUPAC Name 1-piperidin-1-ylethanone
SMILES CC(=O)N1CCCCC1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 mL
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Acetylpiperidine
RTECSNumber TM3975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.