accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 618-80-4

IUPAC Name: 2,6-dichloro-4-nitrophenol
Synonyms: 2,6-dichloro-4-nitrophenol
Molecular Weight (g/mol): 207.99
Molecular Formula: C6H3Cl2NO3
InChi Key: PXSGFTWBZNPNIC-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results