Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-nitrophenol 99.0+%, TCI America™
SDP

Catalog No. D038925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D038925G Supplier TCI America Supplier No. D038925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 618-80-4
Molecular Formula C6H3Cl2NO3
Molecular Weight (g/mol) 207.994
MDL Number MFCD00014715
InChI Key PXSGFTWBZNPNIC-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-nitrophenol, phenol, 2,6-dichloro-4-nitro, 4-nitro-2,6-dichlorophenol, 2,6-dichloro-4-nitro-phenol, 2-6-dichloro-4-nitrophenol, 2,6-dichloro4-nitrophenol, acmc-1b2b6, phenol,6-dichloro-4-nitro, ksc490k9f
PubChem CID 12066
SMILES C1=C(C=C(C(=C1Cl)O)Cl)[N+](=O)[O-]

Specifications

Melting Point 126°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 207.99
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.