accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61832-41-5

IUPAC Name: methyl[(1E)-1-(methylsulfanyl)-2-nitroethenyl]amine
Synonyms: methyl[(1E)-1-(methylsulfanyl)-2-nitroethenyl]amine
Molecular Weight (g/mol): 148.18
Molecular Formula: C4H8N2O2S
InChi Key: YQFHPXZGXNYYLD-ONEGZZNKSA-N
SMILES: CN\C(SC)=C/[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results