Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylamino-1-methylthio-2-nitroethylene 99.0+%, TCI America™
SDP

Catalog No. M130725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M130725G Supplier TCI America Supplier No. M130725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61832-41-5
Molecular Formula C4H8N2O2S
Molecular Weight (g/mol) 148.18
MDL Number MFCD00040994
InChI Key YQFHPXZGXNYYLD-ONEGZZNKSA-N
Synonym n-methyl-1-methylthio-2-nitroethenamine, unii-a9ls0uv4e3, ethenamine, n-methyl-1-methylthio-2-nitro, a9ls0uv4e3, n-methyl-1-methylthio-2-nitro-1-etheneamine, 1-methylamino-1-methylthio-2-nitroethylene, e-n-methyl-1-methylthio-2-nitroethenamine, e-n-methyl-1-methylsulfanyl-2-nitroethenamine
PubChem CID 3035401
IUPAC Name (E)-N-methyl-1-methylsulfanyl-2-nitroethenamine
SMILES CNC(=C[N+](=O)[O-])SC

Specifications

Melting Point 112°C
Color Yellow
Quantity 25 g
Formula Weight 148.18
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylamino-1-methylthio-2-nitroethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.