accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61852-40-2

IUPAC Name: 3-[butyl(phenyl)amino]propanenitrile
Synonyms: 3-[butyl(phenyl)amino]propanenitrile
Molecular Weight (g/mol): 202.3
Molecular Formula: C13H18N2
InChi Key: KMGZOJSHGRNXOC-UHFFFAOYSA-N
SMILES: CCCCN(CCC#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results