Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Cyanoethyl)-N-butylaniline 98.0+%, TCI America™
SDP

Catalog No. C087125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C087125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C087125ML Supplier TCI America Supplier No. C087125ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61852-40-2
Molecular Formula C13H18N2
Molecular Weight (g/mol) 202.301
MDL Number MFCD00019863
InChI Key KMGZOJSHGRNXOC-UHFFFAOYSA-N
Synonym N-Butyl-N-(2-cyanoethyl)aniline, 3-(N-Butyl-N-phenylamino)propionitrile
PubChem CID 112757
IUPAC Name 3-(N-butylanilino)propanenitrile
SMILES CCCCN(CCC#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 mL
Formula Weight 202.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)-N-butylaniline
EINECSNumber (3)-0162
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.