accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFCl(E/Z)

CAS RN 61881-19-4

IUPAC Name: 2,2,2-trifluoro-N-phenylethanecarbonimidoyl chloride
Synonyms: 2,2,2-trifluoro-N-phenylethanecarbonimidoyl chloride
Molecular Weight (g/mol): 207.58
Molecular Formula: C8H5ClF3N
InChi Key: UKKALSJSKAHGTL-UHFFFAOYSA-N
SMILES: FC(F)(F)C(Cl)=NC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results