Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoro-N-phenylacetimidoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T25475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25475G 5 g
T254725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25475G Supplier TCI America Supplier No. T25475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61881-19-4
Molecular Formula C8H5ClF3N
Molecular Weight (g/mol) 207.58
InChI Key UKKALSJSKAHGTL-UHFFFAOYSA-N
PubChem CID 10058869
IUPAC Name 2,2,2-trifluoro-N-phenylethanimidoyl chloride
SMILES C1=CC=C(C=C1)N=C(C(F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 53°C
Quantity 5 g
UN Number 2924
Formula Weight 207.58
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoro-N-phenylacetimidoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.