accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-72-7

IUPAC Name: 4-nitrobenzonitrile
Synonyms: 1-nitro-4-cyanobenzene
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: NKJIFDNZPGLLSH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)