Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitrobenzonitrile

Catalog No. N1200725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1200725G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1200725G Supplier Sigma Aldrich Supplier No. N1200725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phthalylglycyl chloride can be synthesized via reaction of phthalyl-glycine with thionyl chloride.reactant involved in:
  • Synthesis of selenium-containing amino acid analogs with bactericidal and antifungal activity
  • Photoinduced single electron transfer cyclization for preparation of cyclic peptides
  • Arndt-eistert synthesis
  • Mitsunobu and arbuzov-type multicomponent pathway
  • Remote c-h bond functionalization
  • Amido-alkylation for synthesis of benzofuran derivatives

Specifications

CAS 619-72-7
Melting Point 144°C to 147°C (lit.)
Molecular Formula C7H4N2O2
Linear Formula O2NC6H4CN
MDL Number MFCD00007279
Quantity 1 g
Molecular Weight (g/mol) 148.12
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.