CAS RN 619-81-8
IUPAC Name:
cyclohexane-1,4-dicarboxylic acid
Synonyms:
cyclohexane-1,4-dicarboxylic acid
Molecular Weight (g/mol):
172.18
Molecular Formula:
C8H12O4
InChi Key:
PXGZQGDTEZPERC-UHFFFAOYSA-N
SMILES:
OC(=O)C1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
cis-1,4-Cyclohexanedicarboxylic Acid 90.0+%, TCI America™
CAS: 619-81-8 Molecular Formula: C8H12O4 Molecular Weight (g/mol): 172.18 MDL Number: MFCD00064950 InChI Key: PXGZQGDTEZPERC-UHFFFAOYSA-N PubChem CID: 14106 IUPAC Name: cyclohexane-1,4-dicarboxylic acid SMILES: C1CC(CCC1C(=O)O)C(=O)O