Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Cyclohexanedicarboxylic Acid 90.0+%, TCI America™
SDP

Catalog No. C078925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C078925G 25 g
C0789100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C078925G Supplier TCI America Supplier No. C078925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 619-81-8
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00064950
InChI Key PXGZQGDTEZPERC-UHFFFAOYSA-N
PubChem CID 14106
IUPAC Name cyclohexane-1,4-dicarboxylic acid
SMILES C1CC(CCC1C(=O)O)C(=O)O

Specifications

Melting Point 168°C
Color White
Quantity 25 g
Formula Weight 172.18
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,4-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2423&(3)-2446
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.