accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH(Z)

CAS RN 6191-71-5

IUPAC Name: (4Z)-hept-4-en-1-ol
Synonyms: (4Z)-hept-4-en-1-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: CUKAXHVLXKIPKF-ARJAWSKDSA-N
SMILES: CC\C=C/CCCO
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results