accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6191-99-7

IUPAC Name: (2E)-3-ethoxyprop-2-enoyl chloride
Synonyms: (2E)-3-ethoxyprop-2-enoyl chloride
Molecular Weight (g/mol): 134.56
Molecular Formula: C5H7ClO2
InChi Key: SFMFACMIOWQIPR-ONEGZZNKSA-N
SMILES: CCO\C=C\C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results