Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxyacryloyl Chloride 88.0+%, TCI America™
SDP

Catalog No. E10665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10665G Supplier TCI America Supplier No. E10665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6191-99-7
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.559
MDL Number MFCD09029675
InChI Key SFMFACMIOWQIPR-ONEGZZNKSA-N
PubChem CID 5325500
IUPAC Name (E)-3-ethoxyprop-2-enoyl chloride
SMILES CCOC=CC(=O)Cl

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
UN Number 2920
Formula Weight 134.56
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Ethoxyacryloyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.