accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNOOH3CONNNOOOSOOSOOO

CAS RN 61925-55-1

IUPAC Name: bis(2-methoxy-4-nitrobenzene-1-diazonium) naphthalene-1,5-disulfonate
Synonyms: bis(2-methoxy-4-nitrobenzenediazonium) 1,5-naphthalenedisulfonate
Molecular Weight (g/mol): 646.56
Molecular Formula: C24H18N6O12S2
InChi Key: NBJGTSKKWFZFEJ-UHFFFAOYSA-L
SMILES: COC1=CC(=CC=C1[N+]#N)[N+]([O-])=O.COC1=CC(=CC=C1[N+]#N)[N+]([O-])=O.[O-]S(=O)(=O)C1=CC=CC2=C(C=CC=C12)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results