Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Red B Salt 1,5-Naphthalenedisulfonate 97.0+%, TCI America™
SDP

Catalog No. F07511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07511G Supplier TCI America Supplier No. F07511G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61925-55-1
Molecular Formula C24H18N6O12S2
Molecular Weight (g/mol) 646.558
MDL Number MFCD00060048
InChI Key NBJGTSKKWFZFEJ-UHFFFAOYSA-L
Synonym Bis(2-methoxy-4-nitrophenyldiazonium) 1,5-Naphthalenedisulfonate
PubChem CID 151692
IUPAC Name 2-methoxy-4-nitrobenzenediazonium;naphthalene-1,5-disulfonate
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])[N+]#N.COC1=C(C=CC(=C1)[N+](=O)[O-])[N+]#N.C1=CC2=C(C=CC=C2S(=O)(=O)[O-])C(=C1)S(=O)(=O)[O-]

Specifications

Formula Weight 646.56
Percent Purity ≥97.0% (HPLC)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Fast Red B Salt 1,5-Naphthalenedisulfonate [for Biochemical Research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.