CAS RN 61925-78-8
CAS RN 61925-78-8
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-{[(4-methylphenyl)methyl]sulfanyl}propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-{[(4-methylphenyl)methyl]sulfanyl}propanoic acid
Molecular Weight (g/mol):
325.42
Molecular Formula:
C16H23NO4S
InChi Key:
CUNVVZWSABRKAL-UHFFFAOYNA-N
SMILES:
CC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Cys 4-Me-Bzl , Advanced ChemTech
CAS: 61925-78-8 Molecular Formula: C16H23NO4S Molecular Weight (g/mol): 325.42 InChI Key: CUNVVZWSABRKAL-UHFFFAOYNA-N IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-{[(4-methylphenyl)methyl]sulfanyl}propanoic acid SMILES: CC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1