accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 61931-68-8

IUPAC Name: 2-methyl-5-phenyl-1,3-benzoxazole
Synonyms: 2-methyl-5-phenyl-1,3-benzoxazole
Molecular Weight (g/mol): 209.25
Molecular Formula: C14H11NO
InChi Key: CHZDPBVCBBQQRP-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(=CC=C2O1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)